Savant™ SpeedVac™ Modular Vacuum Concentrator, 230 V
Supplier: Thermo Fisher Scientific
SPD120P1-115
76210-556EA 17000 USD
76210-556
Savant™ SpeedVac™ Modular Vacuum Concentrator, 230 V
Evaporators Centrifugal Evaporators
The Thermo Scientific™ Savant™ SpeedVac™ SPD120 vacuum concentrator can be used with a wide variety of solvents and offers resistance to methanol, acetonitrile w/0.1% TFA, and other aggressive solvents used in DNA and biological samples.
  • 230 V model
  • Kit
  • Medium capacity
  • Variety of rotors
  • Glass safety cover
  • PTFE coated aluminum chamber
A medium-capacity, modular-design, the SPD120 vacuum concentrator can also be used for applications where freeze-drying, or lyophilization, is needed.
Ordering information: Supplied as different packages, please see description for details of what is included in specific kits/packages.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (76210-560) is also available, please see separate product page.