ULTIMATE 65™ Double-Knit Polyester, Ultrasonically Sealed Border, Cleanroom Wipes, High-Tech Conversions
Supplier: High-Tech Conversions
ULT65-99
76210-062CS 1050.85 USD
76210-062
ULTIMATE 65™ Double-Knit Polyester, Ultrasonically Sealed Border, Cleanroom Wipes, High-Tech Conversions
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Ultimate 65™ laser sealed-edge heavyweight wipes are extremely absorbent, abrasion resistant, and low in particle emission and ionic levels.
  • Polyester knit
  • Cleanroom packaging
  • 100% polyester cleanroom wipe
The heat of the laser beam cuts and melts each fiber along the edges, reducing the chances of particle emission. Each wipe is laundered in a class 10 cleanroom, individually inspected, and sealed in cleanroom bags.
Packaging: Wipes are packaged 100 per bag, 10 bags per case.