Quad Common Point Ground Cord, Transforming Technologies
Supplier: Transforming Technologies
CP4822
76210-048EA 16.17 USD
76210-048
Quad Common Point Ground Cord, Transforming Technologies
Grounders Grounding Wires
This workstation ground cord features four banana jack receptacles for secure ground connections.
  • Four banana jack receptacles
  • Low profile, molded head
  • Made with durable materials for long life
Made with a low profile, molded head to reduce its footprint, the Quad Common Point Ground (CPG) Cord provides four banana jack receptacles for a ground point for wrist straps, coil cords, matting, or accessories in the area. The low profile, molded head has built-in 10 mm snap for secure worksurface attachment. The 15' black ground cord terminates with a standard 1/8" (#10) ring terminal for a solid connection to ground.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20 and ANSI/ESD S-6.1