Common Point Ground Cord, Transforming Technologies
Supplier: Transforming Technologies
CP1518
76210-040EA 13.35 USD
76210-040
Common Point Ground Cord, Transforming Technologies
Grounders Grounding Wires
Common point ground cords connect multiple aspects of an ESD workstation to ground.
  • Includes one megohm resistor
  • Molded head has a 3/8" male snap for worksurface attachment
Highly durable and reliable, these Common Point Ground (CPG) Cords are equipped with two standard banana jacks, and will ground up to two operators at the same time. The molded head has a built-in male snap for secure worksurface attachment. The 15' black ground cord terminates with a standard 1/8" (#10) ring terminal for a solid connection to ground.
Certifications: Meets or exceeds requirements of ANSI/ESD S20.20 and ANSI/ESD S-6.1