Conductive ESD Trash Can, 44 Gallon, Transforming Technologies
Supplier: Transforming Technologies
WBAS180
76209-970EA 411.13 USD
76209-970
Conductive ESD Trash Can, 44 Gallon, Transforming Technologies
Bins Waste Bins
Conductive ESD trash cans remove the static threat of plastic receptacles that can damage sensitive electronic devices.
  • 44 gallon capacity
  • Permanently conductive; will never lose ESD properties
  • ESD symbol in yellow printed on body for easy identification
  • Typical point-to-point resistance: 10⁴ to 10⁵ ohm
  • Carbon loaded, black conductive polypropylene construction
ESD/anti-static trash cans keep charged waste materials away from sensitive electronic devices and from chemicals that could be ignited by a spark. When grounded, the cans and liners will not hold a static charge and they will contain materials safely. Ideal for cleanrooms, electronics manufacturing, inspection, quality control, and research laboratories. This large and durable trash can is able to withstand manufacturing environments, and will not dent, crush, or chip. Complies with IEC 61340-5-1.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20