Center Snap Common Point Ground Cord System, Transforming Technologies
Supplier: Transforming Technologies
FM2000
76209-886EA 30.62 USD
76209-886
Center Snap Common Point Ground Cord System, Transforming Technologies
Grounders Grounding Wires
This field service ground cord reduces the number of cords to carry with a field service kit.
  • 10' wrist strap cord section
  • 5' ground cord section
  • 10 mm male snap
  • 1 megohm resistor per section
The Common Point Ground Cord System with a unique large center snap and two grounding connections allow for grounding of a wrist strap and work surface. The 10' (extended length) coiled cord section snaps to technicians wrist band; the 5' (extended length) coiled cord section terminates in a standard banana plug which connects to ground either directly or by way of the AC1000 alligator clip (included). Each of the ground cord sections has its own independent one megohm resistor for user safety.
Certifications: Meets or exceeds requirements of ANSI/ESD S20.20 and ANSI/ESD S-6.1