Splicing and maintenance chaps, series 9911, HexArmor
Supplier: HexArmor
9911-S(7) 9911-M(8) 9911-L(9) 9911-XL(10) 9911-XXL(11) 9911-XXXL(12)
76208-112PR 1022.88 USD
76208-112 76208-130 76208-132 76208-134 76208-136 76208-238
Splicing and maintenance chaps, series 9911, HexArmor
Workwear
These HexArmor Series 9911 Chaps protect the body from high-risk hazards on the job site. In addition to high puncture-resistance, these full-length chaps provide ISEA Level 5 cut resistance.
  • Cut, puncture and abrasion resistant, with front-to-back leg coverage
  • Three layers of protection on knee (front only)
  • Reflective tape for visual awareness
  • Launderable
Made with SuperFabric® brand material for some of the highest cut-resistance available in the industry (ANSI/ISEA level A7 and CE level 5).
Certifications: ANSI/ISEA A7, EN 388 4532.