Biox Microtube Rack
Supplier: TWD Scientific
BX90-8009
76208-062CS 96.04 USD
76208-062
Biox Microtube Rack
Racks Tube Racks
Microcentrifuge Tube Racks are made of a high-strength polypropylene that is chemically resistant to alcohols and mild organic solvents. The molded 5×16, 80-Well array is designed for easy tube identification.
  • Polypropylene
  • Easy Mark writing area
  • Equipped with side handles
  • Assorted colors (red, blue, purple, green and yellow)
Microtube racks are designed for use with 1.5 or 2.0 mL microtubes. Racks feature a molded numeric grid for quick identification purposes.