SteelLeather® 5039 Gloves, HexArmor
Supplier: HexArmor
5039-S(7) 5039-M(8) 5039-L(9) 5039-XL(10) 5039-XXL(11) 5039-XXXL(12)
76208-034PR 117.66 USD
76208-034 76208-036 76208-038 76208-040 76208-042 76208-044
SteelLeather® 5039 Gloves, HexArmor
Gloves
The SteelLeather® IX is a heavy-duty solution for times when high cut and puncture–resistance is needed for the entire hand. This glove features split cow leather that will outlast typical leather driver’s gloves.
  • Made with SuperFabric® brand material for some of the highest cut-resistance available in the industry, with ANSI/ISEA level A8 and CE level 5 (full protection coverage)
  • Comfortable split cow leather that will outlast typical leather driver’s gloves
  • May be dry-cleaned for repeated use
  • Hi-vis color scheme for increased visual awareness on the jobsite
SteelLeather® IX 5039 masters safety when high cut and puncture resistance is needed for the entire hand. Heavy-duty split cow leather over SuperFabric® brand material provides ANSI/ISEA level A8 and CE level 5 full protection coverage and superior durability. Gloves may be dry cleaned.