Hercules® 3041 Gloves, HexArmor
Supplier: HexArmor
3041-S(7) 3041-M(8) 3041-L(9) 3041-XL(10) 3041-XXL(11)
76208-024PR 451.11 USD
76208-024 76208-026 76208-028 76208-466 76208-468
Hercules® 3041 Gloves, HexArmor
Gloves
The Hercules® 3041 has a full coverage design and pre-curved shape with a specialized silicon gripping surface on the palm, and the highest levels of needle and cut protection available on the market.
  • Highest needlestick-resistance available
  • Specialized silicon gripping surface on palm
  • Launderable
The Hercules® NSR offers the highest levels of cut-resistance (ANSI/ISEA A9, CE Level 5) and full-hand needlestick protection by layering HexArmor’s exclusively-licensed SuperFabric® brand material. Needlestick protection verified using 25 gauge needles in the ASTM 1342-05 test.