Eyewear Cleaning Fluid, HexArmor
Supplier: HexArmor
14-10006
76207-854EA 10.28 USD
76207-854
Eyewear Cleaning Fluid, HexArmor
Cleaning Agents Lens Cleaning Solutions
Eyewear cleaning fluid, chemically formulated to ensure TruShield® coatings continuously perform to the highest standard.
  • 8 oz. cleaning fluid
  • Suitable for all lenses
  • Fits into wall mount cleaning station
Proprietary coatings are chemically formulated to perform differently than any other coating on the market. Therefore, using standard, non-HexArmor, alcohol-based cleaner on them can cause damage.
For this reason, specific cleaning supplies are available to ensure the TruShield® coatings continuously perform to the highest standard. Using any anti-fog wipes, alcohol-based cleaners, or any other non-HexArmor product on TruShield® lenses can cause damage.