Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Supplier: Agilent Technologies
5188-5217
76204-856EA 6216.44 USD
76204-856 76204-858
Multiple Affinity Removal Column - Mouse 3, Agilent Technologies
Chromatography Columns
The Multiple Affinity Removal LC Column – Mouse 3 is designed as a simple way to remove THREE interfering high-abundant proteins from mouse plasma samples.
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
Caution: For research use only.