Isoamyl alcohol ≥98.5%, clear, colorless liquid ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0219405980 0219405990
76204-324EA 60.52 USD
76204-324 76204-326
Isoamyl alcohol ≥98.5%, clear, colorless liquid ACS
Isoamyl alcohol

  • Grade : ACS
  • Purity : ≥98.5%
  • Key Applications : DNA purification; Conjugation
  • Application Areas : Molecular biology
  • Product Type : Biochemicals
  • Auto Ignition : 350 °C (662 °F)(lit.)
  • Vapor Density : 3.04 (air = 1) (lit.)
  • Vapor Pressure : 2.4 mm Hg at 20 °C 2.37 mm Hg at 25 °C (lit.)
  • Refractive Index : n20/D 1.406(lit.)
  • Presentation : Clear, Colorless Liquid
  • Format : Liquid
  • Water Content : ≤0.5%
  • NOTES : Titrable Acid <0.002 meq/g, Residue after Evaporation <0.003%, Acids and Esters (as amyl acetate) <0.2%, Carbonyl (as HCHO) <0.1%
  • Solubility : Miscible with alcohol, ether, benzene, chloroform, petroleum ether, glacial acetic acid, oils.

Formula: (CH₃)₂CHCH₂CH₂OH
Boiling Pt: 131 °C (268 °F) (lit.)
Melting Pt: –117.2 °C (–179 °F) (lit.)
Density: ∼0.81 g/mL
Flash Pt: 45.5 °C (109.4 °F) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
UN: 1105
ADR: ,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity 98.5%
Water 0.5%
Titrable Acid 0.002 meq/g
Residue after Evaporation 0.003%
Acids and Esters (as amyl acetate) 0.2%
Carbonyl (as HCHO) 0.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR