D-(+)-Galactose (from plant), white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal

02180912.1 02180912.5 0218091201
76204-118EA 57.55 USD
76204-118 76204-120 76204-122
D-(+)-Galactose (from plant), white powder
D(+)-Galactose

D-Galactose is an aldo hexose.


  • Presentation : White Powder
  • Detection Method : FTIR
  • Solubility : Soluble in water (100 mg/mL).

Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Boiling Pt: 527
Storage Temperature: Ambient
MDL Number: MFCD00151230
CAS Number: 59-23-4
Merck Index: 13,04356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adaba957e2b477ca21f11dcfe0e207a3e7fd974a&vid=6ffd412222d8fe5bce2c044ac574ab7c65ee0e92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity >98%
FTIR Conforms to Standard
Melting Point 160 - 170°C
Moisture <1.0%
Optical Rotation
(c=4, water)
+77° ± 5°
Source Plant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR