Glycine, white crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

0219404983 0219404991 0219404992 0219404950
76204-088EA 152.24 USD
76204-088 76204-090 76204-092 76204-322
Glycine, white crystalline powder ACS
Glycine

Glycine is a non-essential amino acid. Only amino acid with no asymmetric carbon. It is a major inhibitory neurotransmitter.


  • Grade : ACS Reagent Grade
  • Assay : ≥98.5%
  • Key Applications : PAGE compound
  • Application Areas : Protein Electrophoresis & Blotting
  • Product Type : Biochemicals
  • Biochemical Category : Amino Acids & Peptides
  • Chemical Class : Amino Acids & Peptides
  • Format : Crystalline Powder
  • pKa :2.35 at 25 °C (Lit.)
  • Heavy Metals : Lead (Pb): ≤0.002%
  • Residue on ignition: ≥0.1%
  • NOTES Chloride (Cl) : ≤0.005%, Sulfate (SO4): ≤0.005%, Ammonium (NH4): ≤0.005%
  • Solubility : Soluble in water (250 mg/mL at 25 °C; 672 mg/mL at 100 °C); slightly soluble in alcohol and ethanol(95%); almost insoluble in ether.

Formula: NH₂CH₂COOH
Melting Pt: 240 °C (dec., lit.)
Density: 1.16 g/mL (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2c080e3c619142b82e0d18a937cd2a089e4c0d7&vid=c2c080e3c619142b82e0d18a937cd2a089e4c0d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.5%
Residue after ignition Not More Than 0.1%
Heavy Metals (as Pb) Not More Than 0.002%
Chloride (Cl) Not More Than 0.005%
Sulfate (SO4) Not More Than 0.005%
Substances darkened by sulfuric acid Passes Test
Hydrolyzable substances Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR