Estradiol, white powder, Cell Culture Grade

Supplier: 0000012207
Warning

02194565.2 0219456501 0219456505
76204-048EA 72.75 USD
76204-048 76204-050 76204-052
Estradiol, white powder, Cell Culture Grade
Estradiol

β-Estradiol is a potent mammalian estrogenic hormone produced by the ovary. Triggers the production of gonadotropins leading to ovulation.


  • Grade : Cell Culture Reagent
  • Suitability : Suitable in Cell Culture
  • Key Applications : Tumorigenicity Studies
  • Product Type : Biochemicals
  • Biochemical Category : Lipids
  • Chemical Class : Lipids
  • xtinction Coefficient : (E1%) EM(280 nm) = 1950 (lit.)
  • Optical Rotation : +76.8 (Concentration = 1, Solvent = Dioxane) (Lit.)
  • Presentation : White Powder
  • Solubility : Almost insoluble in water(50 mg/mL - clear, colorless solution). Freely soluble in alcohol; soluble in acetone, dioxane, other organic solvents; solutions of fixed alkali hydroxides; sparingly soluble in vegetable oils; soluble in methanol:CHCl3 (1:1).


Storage: under nitrogen.

Formula: C₁₈H₂₄O₂
Melting Pt: 173...179 °C
Storage Temperature: Ambient
MDL Number: MFCD00003693
CAS Number: 50-28-2

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard
Melting Point 173 - 179 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR