N,N,N',N'-Tetramethylethylenediamine (TEMED) ≥99% for molecular biology

Supplier: 0000012207
Danger

0219401925 0219401950 0219401980
76203-962EA 64.2 USD
76203-962 76203-964 76204-312
N,N,N',N'-Tetramethylethylenediamine (TEMED) ≥99% for molecular biology
N,N,N',N'-Tetramethylethylenediamine (TEMED)

N,N,N',N'-Tetramethylethylenediamin is a tertiary amine base.


  • Grade : Molecular biology reagent
  • Purity : ~99%
  • Suitability : Suitable for Polyacrylamide Gel Electrophoresis
  • Key Applications : Polyacrylamide gel formation catalyst
  • Application Areas : Nucleic Acid Electrophoresis; Nucleic Acid Sequencing
  • Product Type : Organics
  • Chemical Class : Polar
  • Refractive Index : n20/D 1.4179 (20 °C)(Lit.)
  • Presentation : Liquid
  • pH : 10 - 12 (0.5% aq soln)
  • Detection : Method FTIR
  • Moisture content : ≤2%
  • Solubility : Easily soluble in water (20 gm/mL-clear-cololess solution).


Storage: under nitrogen.

Formula: C₆H₁₆N₂
Boiling Pt: 120…122 °C (lit.)
Melting Pt: –55 °C (lit.)
Density: 0.775 g/mL (20 °C) (lit.)
Flash Pt: 20 °C (68 °F) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00008335
CAS Number: 110-18-9
UN: 2924
ADR: ,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6838f1f5b7d0f69dcc81a8ed247f6c178dad231c&vid=ea361367e9944e4101e38622fdd18b07756eb676&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity >99%
FTIR Conforms to Standard
pH (0.5% aq soln) 10 - 12
Moisture Content <2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR