Anti-P53 Mouse Monoclonal Antibody [clone: PAb122]

Supplier: Enzo Life Sciences
KAM-CC002-D KAM-CC002-F KAM-CC002-J
95040-464EA 258.33 USD
95040-464 95040-466 76203-730
Anti-P53 Mouse Monoclonal Antibody [clone: PAb122]
Antibodies
p53 is a tumor suppressor gene expressed in a wide variety of tissue types and is involved in regulating cell growth, replication, and apoptosis. It binds to Mdm2, SV40 T antigen and human Papilloma virus E6 protein. p53 senses DNA damage and facilitates its repair. Mutation involving p53 is found in a wide variety of malignant tumors, including breast, ovary, bladder, colon, lung, and melanoma.

Type: Primary
Antigen: P53
Clonality: Monoclonal
Clone: PAb122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR