Mini Solvent Pump, Burkle

Supplier: BURKLE INC
5603-4000
76203-678EA 1261.37 USD
76203-678
Mini Solvent Pump, Burkle
Pumps Drum/Transfer Pumps
The Mini solvent pump is a high-quality hand pump for small containers such as bottles, canisters and tanks up to about 10 liters (2.64 gallons).

  • For reliable filling and dosing of solvents and readily flammable liquids
  • Ideal for 1 to 5 l (0.26 to 1.32 gal.) containers
  • Connecting thread GL 45
  • Parts in contact with medium made of PTFE and stainless steel AISI 304 (1.4301), AISI 316L (1.4404)
  • PTFE hose

Simple principle, highly effective–The hand pump works according to the excess pressure principle. An excess pressure is built up in the container with the pump ball, forcing the liquid outward. In this way only the hose, pumping tube and discharge come in contact with the liquid being filled. To ensure safety, a pressure relief valve protects the solvent pump against excessive pressure build-up, for example due to sunshine.

The Mini solvent pump is therefore especially well suited for use with high-purity liquids and readily flammable media such as acetone, isopropanol, ethanol and methanol, etc.

Filling in hazardous zones–The grounding cable included with delivery can easily be connected to the solvent pump with a bunch plug, thus diverting electrostatic charges so that flammable media can be filled safely. Flexible applications–The solvent pump allows for filling from a wide range of container sizes. The flexible PTFE hose that conveys liquid out of the container can be shortened to any length. This ensures ideal residual draining, even from bottles and canisters with a vaulted bottom.

Precise removal of liquids–The self-closing discharge (with auto-stop function) of the Mini solvent pump ensures absolutely precise filling with no dripping at all. Dosing is even easy in very small containers such as test tubes, which present no problems. A hose can optionally be fastened onto the discharge spout (internal diameter 6 mm (0.24")) thus making it possible to fill the container from a greater distance.

The Mini solvent pump is especially compact and very well balanced, which makes it possible to fill without tipping, even from very small containers. Thanks to its sturdy design and high-quality materials, the Mini solvent pump consistently ensures neat and reliable filling throughout its long service life, easily and conveniently.

Accessories information: Adapter available as accessory for S 40, GL 38 and GL 32.

Delivery information: Including grounding cable 2 m (6.56 ft.).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR