Dulbecco's Phosphate-Buffered Saline (DPBS), Himedia®
Supplier: HiMedia
TL1006-500ML
95037-470PK 104.27 USD
95037-470 95037-474 95057-452 76203-532
Dulbecco's Phosphate-Buffered Saline (DPBS), Himedia®
Buffers Cell Culture Buffers
Dulbecco's phosphate buffered saline is used for tissue disaggregation and monolayer dispersal.The presence of calcium and magnesium ions hinder the trypsin activity.
  • Clear, colorless solution
  • pH: 7.20 to 7.80
The solution is used to maintain the medium within physiological pH range and intracellular and extra cellular osmotic balance. Modified with a carbohydrate, such as glucose serves as an energy source for cell metabolism.