Green Caps, MP Biomedicals

Supplier: MP Biomedicals
115067010
76203-086EA 1863.25 USD
76203-086
Green Caps, MP Biomedicals
Closures Test/Sample Tube Closures
Green caps fit EMPTY FASTPREP® tubes and are used in fastprep systems, to hold sample for homogenization.

  • Lysing Matrix D tubes have green caps found in FastRNA™ Pro green kit which are used for isolation of total RNA from plants and animals
  • DNase and RNase-free
  • Store at room temperature(15 - 30 °C)

Use with Empty FastPrep Tubes. Add sample to empty tube, add appropriate Lysing Matrix material and lysing buffer, and apply cap. Process using FastPrep instrument or other bead-beater to lsye, grind or homogenize sample.

Application includes: Sample preparation, sample lysis, nucleic acid isolation and purification, protein isolation, molecular biology, gene expression, microbiological analysis, gene function, gene regulation.

Ordering information: Available separately for those who wish to fill their own lysing matrix tubes.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR