MINI-SPIKE® IV Additive Dispensing Pin, B.Braun
Supplier: B Braun
412012
76202-656CS 221.78 USD
76202-656
MINI-SPIKE® IV Additive Dispensing Pin, B.Braun
Infusions
MINI-SPIKE® IV Additive dispensing pin for preparing and dispensing diluent or additive from multi-dose rubber-stoppered vials
  • Sterile
  • DEHP-free and latex-free
Certifications: CE marked.