Automatic 'One-Touch' Controlled Atmosphere Anaerobic Chamber, Plas-Labs™
Supplier: Plas-Labs
857-OTA
76202-628EA 54517.14 USD
76202-628
Automatic 'One-Touch' Controlled Atmosphere Anaerobic Chamber, Plas-Labs™
Anaerobic Systems
The new 857-OTA One-Touch "Go Anaerobic" chamber allows one simple press of a button to create a strict anaerobic environment. No standing around to manually purge glove box. Simply push the "Go Anaerobic" button on the large, color touch-screen display and the glove box automatically begins the purging sequence. Only 300 liters of gas is needed to achieve anaerobic atmosphere.
  • One-Touch "Go Anaerobic" button
  • One-Touch, on-screen data logging with USB port
  • Large touch screen display
  • Automatic pressure hold function
  • Automatic purging cycles
  • Oxygen analyzer
  • Password protection
  • Two-year warranty
The 857-OTA Anaerobic Chamber includes automatic purging of main chamber and transfer chamber, oxygen analyzer, pressure hold function, and a large easy-to-use touch screen. A drying train system is designed for moisutre and oxygen reduction. Gas Consumption for anaerobic achievement is 300 liters.
Accessories information: No accessories required for standard operation.
Delivery information: Ships on a pallet. Requires loading dock.
Caution: Do not lift by valves. Inspect product immediately upon receipt.