Human Cot DNA, Enzo Life Sciences
Supplier: Enzo Life Sciences
GEN116-0500
76202-436EA
322.64
USD
76202-436
Human Cot DNA, Enzo Life Sciences
Nucleic Acid Reagents
Nucleic Acid Modification and Labelling
Blocks repetitive sequences to prevent nonspecific binding.
- Validated for use in aCGH microarrays and FISH assays
- Decreases background noise to improve signal fidelity
- Prevents nonspecific binding and cross-hybridization in samples
Human Cot DNA blocks repetitive sequences of DNA to suppress the level of cross-hybridization as well as non-specific binding to array surfaces. Utilizing Human Cot DNA in hybridizations increases overall sensitivity and improves experimental results.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)