Anti-CD30 Rabbit Monoclonal Antibody [clone: Ki-1/1505R]

Supplier: Enzo Life Sciences
ABS354-0100
76201-956EA 650.73 USD
76201-956
Anti-CD30 Rabbit Monoclonal Antibody [clone: Ki-1/1505R]
Antibodies
Recombinant monoclonal antibody

  • Recommended Applications: Flow Cytometry, IF, IHC (PS)
  • Species reactivity: Human

Type: Primary
Antigen: CD30
Clonality: Monoclonal
Clone: Ki-1/1505R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR