Anti-RNA polymerase II Mouse Monoclonal Antibody [clone: H14]

Supplier: Enzo Life Sciences
ABS134A-0100
76201-800EA 552.63 USD
76201-800
Anti-RNA polymerase II Mouse Monoclonal Antibody [clone: H14]
Antibodies
Recommended Applications: IF, IP, WB

Species reactivity: Species independent

Type: Primary
Antigen: RNA polymerase II
Clonality: Monoclonal
Clone: H14
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Species independent
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR