ProNex® NGS Library Quant Kit, Promega
Supplier: Promega Corporation
NG1201
76201-682EA 861.03 USD
76201-682
ProNex® NGS Library Quant Kit, Promega
Nucleic Acid Reagents Next Generation Sequencing Reagents
This ProNex® NGS library quant kit contains reagents to determine the concentration of next generation sequencing libraries compatible with Illumina platforms using qPCR.
  • Measure intact, functional library concentration
  • BRYT Green® dye-based qPCR system for maximum reproducibility
  • Ideal for multiplexed pooling of Illumina libraries
Data generated using the ProNex® NGS library quant kit can be used to normalize the DNA concentration for sequencing libraries to the desired concentration prior to multiplex pooling of samples.
Delivery information: Includes ProNex® NGS library Quant 2X master mix (5×1 ml), ProNex® NGS library Quant 10X primer mix (1×1 ml), ProNex® NGS Quant DNA standard (1×25 μl), ProNex® NGS library dilution buffer (2×150 ml) and nuclease-free water (2×1250 μl).
Caution: For research use only. Not for use in diagnostic procedures. Storage Temperature –30...–10 °C