CTAB Buffer, Promega
Supplier: Promega Corporation
MC1411
76201-566EA 57.55 USD
76201-566
CTAB Buffer, Promega
Buffers Extraction Buffers
Lysis buffer for use with the Maxwell® RSC PureFood GMO and Authentication Kit.