Quantus™ NGS Starter Package, Promega
Supplier: Promega Corporation
E5150
76201-548EA 2814.74 USD
76201-548
Quantus™ NGS Starter Package, Promega
Nucleic Acid Reagents Next Generation Sequencing Reagents
Highly sensitive and easy-to-use DNA quantitation for your NGS applications all in one discounted bundle. QuantiFluor® dyes are optimized for high sensitivity, broad dynamic range and target specificity on the Quantus™ Fluorometer.
  • Use for next-generation sequencing
  • Measure low dsDNA
  • Integrated instrumentation and assay
  • Add-and-read format makes measuring low concentrations of dsDNA simple - no dilutions, no extra tubes
  • Significantly increased sensitivity over absorbance at 260 nm (NanoDrop spectrophotometer) for those samples that are low in concentration
  • Expect high specificity to dsDNA with minimal binding to ssDNA, RNA, protein and interfering compounds
The Quantus™ fluorometer is a compact and easy-to-operate instrument designed for highly sensitive fluorescent detection of nucleic acids. The Quantus™ fluorometer is optimized with preprogrammed settings for QuantiFluor® Dye Systems (QuantiFluor® dsDNA, RNA, ssDNA Systems) for nucleic acid quantitation and offers the flexibility to create customized methods and quantitation settings for other dyes.
QuantiFluor® ONE dsDNA system provides a fluorescent double-stranded DNA-binding dye in an add-and-read format for both dye and standard, simplifying DNA quantitation and speeding up the workflow. It's as easy to use as NanoDrop® absorbance-based methods but much more sensitive for low-concentration samples.
Delivery information: Package includes Quantus™ fluorometer, QuantiFluor® ONE dsDNA system (1×500 reactions), 0.5 ml PCR tubes (10×50/pack).
Caution: For Research Use Only. Not for Use in Diagnostic Procedures. Store QuantiFluor® ONE dsDNA Dye and QuantiFluor® ONE Lambda DNA at –30...+10 °C. Store 1X TE Buffer at –30...+30 °C