Glycine ≥99.0% for molecular biology

Supplier: Promega Corporation

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

H5073
PAH5071EA 77.75 USD
PAH5071 76201-024
Glycine ≥99.0% for molecular biology
Glycine

Glycine is an amino acid used in the preparation of some electrophoresis buffers.


  • Quality tested and certified free of DNase, RNase and protease activity


Storage: 15 °C to 30 °C.

Formula: NH₂CH₂COOH
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99.0%
Iron ≤10ppm
A260 at 1M ≤0.05
A280 at 1M ≤0.05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR