Kinesis® TELOS® QuEChERS Extraction Kits, Cole-Parmer®
Supplier: Antyila Scientific
Quickly and economically extract the required analytes into solution.
- Salt pack helps transfer salt to the centrifuge tube quickly and easily with minimal waste
- Break up complex sample types with homogenizer balls
TELOS® QuEChERS extraction kits offer the first stage cleanup for many sample types. This initial crude extraction and partitioning step uses organic solvent and salt solution to extract the required analytes into solution. Select from extraction kits without a tube, kits with a 50 ml centrifuge tube, or kits with a tube and homogenizer beads. Beads aid in breaking up more complex sample types.
Certifications: Conform with documented AOAC 2007.01 and EN 15662 protocols.