SterEM™ Sabdex Agar with Lecithin and Tween® 80, USP, Irradiated
Supplier: HARDY DIAGNOSTICS
W595
76200-290PK
82.77
USD
76200-290
SterEM™ Sabdex Agar with Lecithin and Tween® 80, USP, Irradiated
Media for Microbiology
Dehydrated Media for Microbiology
Sterile plates for environmental monitoring.
- Room temperature storage
- Improved moisture control packaging format
- Optically clear polystyrene - For distortion free viewing
- Heavy-Duty - Reduces breakage during shipping
- Stacking rings - Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding
- Sterile by Irradiation - Validated for Sterility Assurance Level (SAL) of 10⁻⁵
- Triple Bagged - Two nylon inner bags. One outer Mylar(R) bag that is heat sealed
- Outer bag is VHP impermeable
- Certificate of Analysis - Available at the Hardy Diagnostics website 24/7
Irradiated Sabouraud Dextrose (SabDex) agar with lecithin and Tween® 80 is recommended for the detection and enumeration of yeasts and fungi from environmental sources in a clean room setting. The plates are triple bagged and sterilized by irradiation to promote a higher sterility assurance level.
Certifications: Manufactured in a licensed cGMP facility with a quality management system certified to ISO 13485.