Lab-Elite™ Certified Reference Materials (CRM), KWIK-STIK, Microbiologics
Supplier: MICROBIOLOGICS, INC.
01056-CRM 0111-CRM 0261-CRM 0262-CRM 0817-CRM 0890-CRM 0902-CRM 0947-CRM 0968-CRM 0317-CRM 0421-CRM 0441-CRM 0604-CRM 0687-CRM 0697-CRM 0727-CRM 01045-CRM 0356-CRM 0789-CRM 0805-CRM 0121-CRM 0345-CRM
76199-934PK 102.71 USD
76199-934 76199-936 76199-942 76199-944 76199-952 76199-954 76199-956 76200-006 76200-008 76200-012 76200-018 76200-020 76200-024 76200-028 76200-030 76200-032 76200-130 76200-138 76200-140 76200-142 76200-194 76200-204
Lab-Elite™ Certified Reference Materials (CRM), KWIK-STIK, Microbiologics
Controls and Calibration Verification Materials
Our Lab-Elite CRM is designed to help those labs meet the requirements of ISO 17025 standards. They are just one passage from the reference culture and are delivered in our signature KWIK-STIK format.
  • Traceable to reference culture
  • Refrigerated storage is easy and economical
  • Ready-to-use format to save you time and money
  • Strains are one passage from reference culture
  • Technical Support experts available for guidance
This standard specifies that "reference materials shall, where possible, be traceable to SI units of measurements, or to Certified Reference Materials". As a Certified Reference Material, Lab-Elite comes with a comprehensive Certificate of Analysis detailing the strains identity, characteristics and standard deviation. Qualitative QC microorganisms one passage from the reference strains for ultimate authenticity.