Anti-IgG Hamster Polyclonal Antibody

Supplier: Immunoreagents
HA-003-ZBIO
76199-650EA 125.61 USD
76199-650
Anti-IgG Hamster Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Hamster
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR