SCREEN-WELL® Cancer Library, Enzo Life Sciences
Supplier: Enzo Life Sciences
LIB102-0100
76196-694EA 7380.39 USD
76196-694
SCREEN-WELL® Cancer Library, Enzo Life Sciences
Standards Biological Standards
Essential standards for cancer inhibitor screening.
This Cancer Library is a collection of 275 compounds that can be used for cancer inhibitor screening and assay development. The library contains small molecules affecting mTOR, aurora kinases, tyrosine kinases, PI3K, and HDAC, as well as many structurally and mechanistically different compound classes. This library is an essential tool for cancer inhibitor screening and drug development. The compounds are dissolved in DMSO at 10 mM and aliquoted into 96-well deep-well plates at 100 µL per well.
Ordering information: Contains 275 structurally and mechanistically different compounds, as well as nontoxic controls.