IL-6 High Sensitivity Controls, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-KIT178-CTL
76196-692EA 163.5 USD
76196-692
IL-6 High Sensitivity Controls, Enzo Life Sciences
Standards Biological Standards
IL-6 Controls for use with IL-6 High Sensitivity ELISA Kit.
  • Lyophilized set of controls
This product is a set of quantitative controls for the determination of IL-6 concentration in biological matrices, such as serum and plasma. The concentrations have been determined using the IL-6 (human) high sensitivity ELISA kit. The controls were lyophilized in porcine serum.
Ordering information: Contains 1 high, 1 medium, and 1 low control.