HIGHDEF® Green AP Chromogen/Substrate, Enzo Life Sciences
Supplier: Enzo Life Sciences
ACC130-0030
76196-688EA 317.19 USD
76196-688
HIGHDEF® Green AP Chromogen/Substrate, Enzo Life Sciences
Assays Immunological Assays
High definition IHC chromogen producing brilliant green color for AP-activated development.
This chromogen or substrate is intended to be used in conjunction with alkaline phosphatase (AP)-based immunostaining or in-situ hybridization systems.
Ordering information: Contains HIGHDEF® Green AP Chromogen, HIGHDEF® Green AP Substrate.