SATWipes® Replacement Canister
Supplier: Contec
CSCN0100
76196-618CS 215.42 USD
76196-618
SATWipes® Replacement Canister
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
A replacement canister for the SATWipes® 6×9" rolls saturated with IPA greater than 70%.
  • Canister features a reinforced snap-close lid