Czapek Dox Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
W119
76196-444PK 184.13 USD
76196-444
Czapek Dox Agar, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
Accommodates deeper fills. Used for the cultivation of saprophytic fungi and soil bacteria capable of utilizing inorganic nitrogen.
  • Neutral pH