NEB Tube Opener, New England Biolabs
Supplier: New England Biolabs (NEB)
C1008S
76196-412EA 25 USD
76196-412
NEB Tube Opener, New England Biolabs
Crimpers, Cappers and Decappers
The NEB tube opener can be used to open a wide variety of microcentrifuge tubes.
Can be used for snap-on caps or screw-on caps.