MidRange PFG Marker, New England Biolabs
Supplier: New England Biolabs (NEB)
N0342S
76196-402EA 184 USD
76196-402
MidRange PFG Marker, New England Biolabs
Electrophoresis Reagents Molecular Weight Markers
MidRange PFG Marker consists of concatemers of DNA isolated from the bacteriophage (cI857 ind1 Sam7) mixed with XhoI digested DNA embedded in 1% LMP agarose and supplied in a GelSyringe dispenser.
  • Suitable for pulsed field gel electrophoresis
  • Size range: 15 to 291 kb Embedded in 1% Low Melting Point (LMP) agarose
  • Supplied in a GelSyringe™ dispenser
  • Suitable for 50 gel lanes
XhoI produces fragments of 15.0 and 33.5 kb. These fragments anneal to and form concatemers with intact DNA. It is designed for use as a size marker for pulsed field gel electrophoresis (PFG).