Vein Pick Catheter Introducer, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
VP-10
76193-114EA 224.17 USD
76193-114
Vein Pick Catheter Introducer, SAI Infusion Technologies
Syringes Syringes, Medical
This simple and effective tool holds the vessel open during a cut-down catheter insertion procedure.
  • Tapered tip for easy insertion and vessel dilation
  • Tunnel feature makes it easy to direct the catheter into the vessel
  • Supplied sterile
Ideal for use with rats, it can also be used for large animal surgeries. Simply slide the catheter through the groove on the underside of the vein pick and into the vessel for a stress-free catheter implantation.
Caution: This product is designed for laboratory animal use only.