Econo™ Joseph Hook, Floor Grade, Sklar®
Supplier: Sklar
21-390 21-392 21-394
76192-468BX 172.92 USD
76192-468 76192-470 76192-472
Econo™ Joseph Hook, Floor Grade, Sklar®
Retractors
Econo™ Joseph Skin Hook is a handheld retractor commonly used for holding back the edges of skin.
  • With industry standard
  • With five major surgical instruments lines
  • Delivers superior precision for any surgical procedure
It is commonly used during intranasal and pharyngeal procedures. It is available with a single or dual prong, sharp hook. The handle is often round and knurled and it tapers to the hook end. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.