Econo™ Metzenbaum-Lahey Dissecting Scissors, Floor Grade, Sklar®
Supplier: Sklar
21-344
76192-452BX 91.39 USD
76192-452
Econo™ Metzenbaum-Lahey Dissecting Scissors, Floor Grade, Sklar®
Scissors
Econo™ Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Econo™ Metzenbaum-Lahey Dissecting Scissors are not recommended for cutting sutures, drains, or heavy tissue. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.