Stille Barraya Forceps, OR Grade, Sklar®
Supplier: Sklar
19-1990
76192-320EA 101.87 USD
76192-320
Stille Barraya Forceps, OR Grade, Sklar®
Forceps
Sklar® Stille-Barraya Forceps are long thumb style forceps commonly used for grasping organs or dissecting tissue in the stomach and intestines.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard