Dressing Forceps, Fluted Handle, OR Grade, Sklar®
Supplier: Sklar
19-1340 19-1341 19-1342 19-1343 19-1345 19-1347
76192-260EA 51.94 USD
76192-260 76192-262 76192-264 76192-266 76192-268 76192-270 76192-272
Dressing Forceps, Fluted Handle, OR Grade, Sklar®
Forceps
Sklar's® Fluted Handle Dressing Forceps are used in a wide variety of procedures.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar's® fluted handle dressing forceps are used in a wide variety of procedures for holding dressing materials such as gauze, changing dresings, or packing wounds. The fluted handle offers an ergonomic non-slip grip. This product has serrated blunt tips.