Birkett Hemostatic Forceps, OR Grade, Sklar®
Supplier: Sklar
17-3210 17-3211 17-3212 17-3213
76192-096EA 139.61 USD
76192-096 76192-226 76192-228 76192-230
Birkett Hemostatic Forceps, OR Grade, Sklar®
Forceps
Birkett Hemostatic Forceps are slender, ratcheted, finger ring forceps.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Sklar® Birkett Hemostatic Forceps are used for clamping vessels and controlling blood flow. They are similar to the Moynihan hemostat, with fully serrated jaws and commonly used in procedures in the abdominal area. However, the Birkett hemostat has slightly larger jaws and is often slightly longer.