Sklarlite™ Sklarhone™ Metzenbaum-Nelson Bariatric Scissors, OR Grade, Sklar®
Supplier: Sklar
15-9119
76192-028EA 324.24 USD
76192-028
Sklarlite™ Sklarhone™ Metzenbaum-Nelson Bariatric Scissors, OR Grade, Sklar®
Scissors
Sklarlite™ Sklarhone™ Metzenbaum-Nelson Bariatric Scissors are long dissecting scissors most commonly used in Bariatric Surgeries such as gastric bypass, gastric banding, and sleeve gastrectomy. They are also used in Nephrectomy, Cystectomy, and Prostatectomy procedures to dissect and cut tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Metzenbaum-Nelson scissors tend to be longer than standard Metzenbaum scissors, making them ideal for use in thoracic procedures as well. These scissors are not recommended for cutting sutures or drains. The Sklarlite™ Sklarhone™ combination provides the surgeon with a lightweight, precision instrument to make the sharp incisions of a knife with the cutting ability of scissors, all while having greater control and experiencing less fatigue. This instrument is identified by dual black finger rings.