Sprague-Rappaport Stethoscope, Sklar®
Supplier: Avantor
06-2320
76191-628EA 62.54 USD
76191-628 76191-632 76191-634 76191-638 76191-640 76191-642 76191-644 76191-646
Sprague-Rappaport Stethoscope, Sklar®
Stethoscopes
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Sprague-Rappaport Diagnostic Stethoscope features thick, dual tubing to pick up even very faint sounds. This 5-in-1 stethoscope can be used on adult, pediatric, and infant patients. It comes with two diaphragms, three bells, and two additional ear pieces. The large diaphragm and the large bell are used withMore Product Information