Schink Dermatome, OR-Grade, Sklar®
Supplier: Sklar
06-6300
76191-440EA 3748.57 USD
76191-440
Schink Dermatome, OR-Grade, Sklar®
Knives
Sklar's® Schink Dermatome is a dermal instrument also known as a skin grafting knife. This instrument is used to slice thin portions of skin from one part of the body to be used in grafting another, injured part of the body.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Severe burns and similar traumas are what commonly warrants the use of this instrument. Dermatomes can be both manually and electrically operated. This Sklar Dermatome is a manual instrument. Straight, Sharp, Complete Instrument with two Skin Fixing Plates and 1 Replaceable Blade. Adjustable from 0.1 to 0.2 mm.